Ethyl-3-acetylbenzoate 98%

Lieferant: Apollo Scientific

OR952486-1G OR952486-5G
APOSOR952486-1GEA 0 EUR
APOSOR952486-1G APOSOR952486-5G
Ethyl-3-acetylbenzoate 98%
Ethyl-3-acetylbenzoate
Formel: C₁₁H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02261283
CAS-Nummer: 37847-24-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR