7-Bromo-3-methyl-1H-indole ≥98%

Lieferant: Apollo Scientific

OR952575-100MG
APOSOR952575-100MGEA 188 EUR
APOSOR952575-100MG
7-Bromo-3-methyl-1H-indole ≥98%
7-Bromo-3-methyl-1H-indole
Formel: C₉H₈BrN
Lagertemperatur: Raumtemperatur
MDL: MFCD07799775
CAS-Nummer: 86915-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR