3-Pyrrol-1-ylpropionic acid 97%

Lieferant: Apollo Scientific
Achtung

OR952599-10G OR952599-1G OR952599-5G
APOSOR952599-10GEA 864 EUR
APOSOR952599-10G APOSOR952599-1G APOSOR952599-5G
3-Pyrrol-1-ylpropionic acid 97%
3-Pyrrol-1-ylpropionic acid
Formel: C₇H₉NO₂
Schmelzpunkt: 59…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03844729
CAS-Nummer: 89059-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR