4-Methoxy-3-nitropyridin

Lieferant: Apollo Scientific
Achtung

OR9526-25G
APOSOR9526-25GEA 63.8 EUR
APOSOR9526-25G
4-Methoxy-3-nitropyridin
4-Methoxy-3-nitropyridine
Formel: C₆H₆N₂O₃
Molecular Weight: 154.13 g/mol
Schmelzpunkt: 72…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00209661
CAS-Nummer: 31872-62-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR