1-Ethyl-1H-pyrazole-3-carboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 1-Ethylpyrazole-3-carboxylic Acid

OR952629-5G
APOSOR952629-5GEA 136 EUR
APOSOR952629-5G
1-Ethyl-1H-pyrazole-3-carboxylic acid 98%
1-Ethyl-1H-pyrazole-3-carboxylic acid
Formel: C₆H₈N₂O₂
Molecular Weight: 140.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02090832
CAS-Nummer: 400755-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR