2,3-Dihydro-[1,4]dioxino[2,3-b]pyridin-7-amine 95%

Lieferant: Apollo Scientific
Achtung

OR952663-250MG
APOSOR952663-250MGEA 426 EUR
APOSOR952663-250MG
2,3-Dihydro-[1,4]dioxino[2,3-b]pyridin-7-amine 95%
2,3-Dihydro[1,4]dioxino[2,3-b]pyridin-7-amine
Formel: C₇H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18374085
CAS-Nummer: 1261365-47-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR