Ethyl thiomorpholine-2-carboxylate 95%
Lieferant: Apollo Scientific
OR952686-1G
APOSOR952686-1GEA
719
EUR
APOSOR952686-1G
Ethyl thiomorpholine-2-carboxylate 95%
Ethyl thiomorpholine-2-carboxylate
Formel:
C₇H₁₃NO₂S Molecular Weight: 175,249 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20134212 CAS-Nummer: 152009-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...