(2-Formyl-4-nitrophenoxy)acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Formyl-4-nitrophenoxy)ethanoic acid, 2-(2-Formyl-4-nitrophenoxy)acetic acid, 2-(Carboxymethoxy)-5-nitrobenzaldehyde

OR9527-1G OR9527-5G OR9527-25G
APOSOR9527-1GEA 21.9 EUR
APOSOR9527-1G APOSOR9527-5G APOSOR9527-25G
(2-Formyl-4-nitrophenoxy)acetic acid
(2-Formyl-4-nitrophenoxy)acetic acid
Formel: C₉H₇NO₆
Molecular Weight: 225.155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024502
CAS-Nummer: 6965-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR