2-Anilinopyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Phenylpyridin-2-amine
OR952715-25G
APOSOR952715-25GEA
183
EUR
APOSOR952715-25G
2-Anilinopyridine 98%
2-Anilinopyridine
Formel:
C₁₁H₁₀N₂ Molecular Weight: 170.215 g/mol Siedepunkt: 205…206 °C/7mmHg Schmelzpunkt: 106…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006248 CAS-Nummer: 6631-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...