N-Cyclopropyl-N-isopropylamine hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
N-Cyclopropyl-n-isopropylamine, HCl
OR952717-1G
APOSOR952717-1GEA
371
EUR
APOSOR952717-1G
N-Cyclopropyl-N-isopropylamine hydrochloride 95%
N-Cyclopropyl-N-isopropylamine hydrochloride
Formel:
C₆H₁₃N·ClH Molecular Weight: 135,635 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09863848 CAS-Nummer: 246257-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...