Acetylamino-cyclopentyl-acetic acid 95%

Lieferant: Apollo Scientific

OR952789-1G
APOSOR952789-1GEA 0 EUR
APOSOR952789-1G
Acetylamino-cyclopentyl-acetic acid 95%
Acetylamino-cyclopentyl-acetic acid
Formel: C₉H₁₅NO₃
Molecular Weight: 185.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06796733
CAS-Nummer: 2521-83-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR