6-Chloro-1,3-benzoxazol-2-amine 97%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-6-chlorobenzoxazole
OR952811-5G
APOSOR952811-5GEA
95.7
EUR
APOSOR952811-5G
6-Chloro-1,3-benzoxazol-2-amine 97%
6-Chloro-1,3-benzoxazol-2-amine
Formel:
C₇H₅ClN₂O Molecular Weight: 168.58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01664218 CAS-Nummer: 52112-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...