Benzooxazole-2-carboxylic acid methyl ester 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl benzo[d]oxazole-2-carboxylate
OR952861-1G
APOSOR952861-1GEA
94.4
EUR
APOSOR952861-1G
Benzooxazole-2-carboxylic acid methyl ester 95%
Benzooxazole-2-carboxylic acid methyl ester
Formel:
C₉H₇NO₃ Molecular Weight: 177,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09701312 CAS-Nummer: 27383-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...