Isoquinoline-4-carbothioic acid amide 95%

Lieferant: Apollo Scientific

Synonyme: Isoquinoline-4-carbothioamide

OR952895-1G OR952895-250MG
APOSOR952895-1GEA 243 EUR
APOSOR952895-1G APOSOR952895-250MG
Isoquinoline-4-carbothioic acid amide 95%
Isoquinoline-4-carbothioic acid amide
Formel: C₁₀H₈N₂S
Molecular Weight: 188.249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738908
CAS-Nummer: 435271-32-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR