4-Acetamido-3-ethoxynitrobenzene

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(2-Ethoxy-4-nitrophenyl)acetamide

OR9529-1G OR9529-5G OR9529-25G
APOSOR9529-1GEA 21.9 EUR
APOSOR9529-1G APOSOR9529-5G APOSOR9529-25G
4-Acetamido-3-ethoxynitrobenzene
4-Acetamido-3-ethoxynitrobenzene
Formel: C₁₀H₁₂N₂O₄
Molecular Weight: 224.213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00994990
CAS-Nummer: 116496-76-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR