(1-Phenylpiperidin-4-yl)methanol 95%
Lieferant: Apollo Scientific
OR952959-1G
APOSOR952959-1GEA
271
EUR
APOSOR952959-1G
(1-Phenylpiperidin-4-yl)methanol 95%
(1-Phenylpiperidin-4-yl)methanol
Formel:
C₁₂H₁₇NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410644 CAS-Nummer: 697306-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...