5-Methoxyisoxazol-3-amine 97%

Lieferant: Apollo Scientific

OR952969-1G OR952969-250MG OR952969-5G
APOSOR952969-1GEA 0 EUR
APOSOR952969-1G APOSOR952969-250MG APOSOR952969-5G
5-Methoxyisoxazol-3-amine 97%
5-Methoxyisoxazol-3-amine
Formel: C4H6N2O2
Lagertemperatur: Raumtemperatur
CAS-Nummer: 32326-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR