2-(4-Piperidyl)-2-propanol 96%
Lieferant: Apollo Scientific
Synonyme:
2-(4-Piperidyl)isopropyl Alcohol
OR953011-25G
OR953011-5G
APOSOR953011-25GEA
330
EUR
APOSOR953011-25G
APOSOR953011-5G
2-(4-Piperidyl)-2-propanol 96%
2-(4-Piperidyl)-2-propanol
Formel:
C₈H₁₇NO Molecular Weight: 143.23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797159 CAS-Nummer: 22990-34-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...