cis-4-Methyl-cyclohexylamine hydrochloride 95%

Lieferant: Apollo Scientific

OR953018-100MG OR953018-1G OR953018-250MG
APOSOR953018-100MGEA 0 EUR
APOSOR953018-100MG APOSOR953018-1G APOSOR953018-250MG
cis-4-Methyl-cyclohexylamine hydrochloride 95%
cis-4-Methyl-cyclohexylamine hydrochloride
Formel: C₇H₁₅N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD09991703
CAS-Nummer: 33483-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR