Methyl-3-nitrophenylacetate 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-Nitrophenylacetate

OR953022-5G
APOSOR953022-5GEA 105 EUR
APOSOR953022-5G
Methyl-3-nitrophenylacetate 97%
Methyl-3-nitrophenylacetate
Formel: C₉H₉NO₄
Molecular Weight: 195.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669939
CAS-Nummer: 10268-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR