2-(Cyclopentyloxy)ethylamine 95%

Lieferant: Apollo Scientific

OR953041-1G
APOSOR953041-1GEA 203 EUR
APOSOR953041-1G
2-(Cyclopentyloxy)ethylamine 95%
2-(Cyclopentyloxy)ethylamine
Formel: C₇H₁₅NO
Molecular Weight: 129.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09730285
CAS-Nummer: 933732-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR