Methyl thiazole-2-carboxylate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-thiazolecarboxylate
OR953064-5G
APOSOR953064-5GEA
89.9
EUR
APOSOR953064-5G
Methyl thiazole-2-carboxylate 98%
Methyl thiazole-2-carboxylate
Formel:
C₅H₅NO₂S Molecular Weight: 143.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18075580 CAS-Nummer: 55842-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...