Methylpyrazolo[1,5-a]pyridin-2-carboxylat 95%

Lieferant: Apollo Scientific

Synonyme: Pyrazolo[1,5-a]pyridine-2-carboxylic acid methyl ester

OR953065-1G
APOSOR953065-1GEA 235 EUR
APOSOR953065-1G
Methylpyrazolo[1,5-a]pyridin-2-carboxylat 95%
Methylpyrazolo[1,5-a]pyridin-2-carboxylat
Formel: C₉H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07367039
CAS-Nummer: 151831-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR