Ethyl-4-aminopyrimidine-5-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-aminopyrimidine-5-carboxylate

OR953105-1G
APOSOR953105-1GEA 49.3 EUR
APOSOR953105-1G
Ethyl-4-aminopyrimidine-5-carboxylate 97%
Ethyl-4-aminopyrimidine-5-carboxylate
Formel: C₇H₉N₃O₂
Molecular Weight: 167,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01028678
CAS-Nummer: 65195-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR