1-Methyl-1H-indol-6-amine 96%
Lieferant: Apollo Scientific
Synonyme:
6-Amino-1-methylindole
OR953134-1G
APOSOR953134-1GEA
155
EUR
APOSOR953134-1G
1-Methyl-1H-indol-6-amine 96%
1-Methyl-1H-indol-6-amine
Formel:
C₉H₁₀N₂ Molecular Weight: 146,189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10000822 CAS-Nummer: 135855-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...