Dimethyl (R)-(+)-methylsuccinate 97%

Lieferant: Apollo Scientific

Synonyme: Dimethyl (2R)-2-methylbutanedioate

OR953141-1G
APOSOR953141-1GEA 174 EUR
APOSOR953141-1G
Dimethyl (R)-(+)-methylsuccinate 97%
Dimethyl (R)-(+)-methylsuccinate
Formel: C₇H₁₂O₄
Molecular Weight: 160,17 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 22644-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR