2-Methoxy-5-methylnicotinonitrile 98%

Lieferant: Apollo Scientific

OR953188-1G
APOSOR953188-1GEA 632 EUR
APOSOR953188-1G
2-Methoxy-5-methylnicotinonitrile 98%
2-Methoxy-5-methylnicotinonitrile
Formel: C₈H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD13563068
CAS-Nummer: 149379-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR