2,6-Dichlor-α-methylbenzylalkohol 95%

Lieferant: Apollo Scientific

OR953192-5G
APOSOR953192-5GEA 55.1 EUR
APOSOR953192-5G
2,6-Dichlor-α-methylbenzylalkohol 95%
2,6-Dichlor-α-methylbenzylalkohol
Formel: C₈H₈Cl₂O
Molecular Weight: 191.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016853
CAS-Nummer: 53066-19-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR