2,5-Dichlor-α-methylbenzylalkohol 95%
Lieferant: Apollo Scientific
OR953205-25G
APOSOR953205-25GEA
111
EUR
APOSOR953205-25G
2,5-Dichlor-α-methylbenzylalkohol 95%
2,5-Dichlor-α-methylbenzylalkohol
Formel:
C₈H₈Cl₂O Molecular Weight: 191.06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021861 CAS-Nummer: 1475-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...