Methyl 5-(aminomethyl)-2-methoxybenzoate hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR953223-1G OR953223-250MG
APOSOR953223-1GEA 835 EUR
APOSOR953223-1G APOSOR953223-250MG
Methyl 5-(aminomethyl)-2-methoxybenzoate hydrochloride 95%
Formel: C₁₀H₁₃NO₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD07285352
CAS-Nummer: 1049782-92-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR