Benzyl(furan-3-ylmethyl)amine 97%

Lieferant: Apollo Scientific

OR953246-1G
APOSOR953246-1GEA 435 EUR
APOSOR953246-1G
Benzyl(furan-3-ylmethyl)amine 97%
Benzyl(furan-3-ylmethyl)amine
Formel: C₁₂H₁₃NO
Molecular Weight: 187,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12546988
CAS-Nummer: 179057-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR