1H-Imidazo[4,5-b]pyridin-2(3H)-one 98%

Lieferant: Apollo Scientific

Synonyme: 1,3-Dihydro-2H-imidazo[4,5-b]pyridin-2-one

OR953324-1G
APOSOR953324-1GEA 132 EUR
APOSOR953324-1G
1H-Imidazo[4,5-b]pyridin-2(3H)-one 98%
1H-Imidazo[4,5-b]pyridin-2(3H)-one
Formel: C₆H₅N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00464845
CAS-Nummer: 16328-62-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR