1,4-Dioxaspiro[4.5]decane-8-carbonitrile 95%

Lieferant: Apollo Scientific

OR953326-5G
APOSOR953326-5GEA 121 EUR
APOSOR953326-5G
1,4-Dioxaspiro[4.5]decane-8-carbonitrile 95%
1,4-Dioxaspiro[4.5]decane-8-carbonitrile
Formel: C₉H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09037879
CAS-Nummer: 69947-09-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR