4-(Methylamino)cyclohexanol 97%
Lieferant: Apollo Scientific
OR953332-1G
OR953332-5G
APOSOR953332-1GEA
219
EUR
APOSOR953332-1G
APOSOR953332-5G
4-(Methylamino)cyclohexanol 97%
4-(Methylamino)cyclohexanol
Formel:
C₇H₁₅NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09909445 CAS-Nummer: 2987-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...