4-(1H-Imidazol-2-yl)benzoic acid 96%

Lieferant: Apollo Scientific

OR953354-5G
APOSOR953354-5GEA 95.7 EUR
APOSOR953354-5G
4-(1H-Imidazol-2-yl)benzoic acid 96%
4-(1H-Imidazol-2-yl)benzoic acid
Formel: C₁₀H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00456057
CAS-Nummer: 108035-45-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR