Methyl-3-methyl-2-thiophencarboxylat 96%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-methylthiophene-2-carboxylate
OR953363-25G
APOSOR953363-25GEA
85.7
EUR
APOSOR953363-25G
Methyl-3-methyl-2-thiophencarboxylat 96%
Methyl-3-methyl-2-thiophencarboxylat
Formel:
C₇H₈O₂S Molecular Weight: 156.20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234332 CAS-Nummer: 81452-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...