2-Phenoxybenzonitril 98%

Lieferant: Apollo Scientific

OR953387-1G
APOSOR953387-1GEA 29 EUR
APOSOR953387-1G
2-Phenoxybenzonitril 98%
2-Phenoxybenzonitril
Formel: C₁₃H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD08271960
CAS-Nummer: 6476-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR