Ethyl-4-acetylbenzoate 95%

Lieferant: Apollo Scientific

OR953424-1G OR953424-5G
APOSOR953424-1GEA 0 EUR
APOSOR953424-1G APOSOR953424-5G
Ethyl-4-acetylbenzoate 95%
Ethyl-4-acetylbenzoate
Formel: C₁₁H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00013241
CAS-Nummer: 38430-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR