(1-Phenyl-1H-pyrazol-4-yl)methanol 97%
Lieferant: Apollo Scientific
OR953502-1G
APOSOR953502-1GEA
109
EUR
APOSOR953502-1G
(1-Phenyl-1H-pyrazol-4-yl)methanol 97%
(1-Phenyl-1H-pyrazol-4-yl)methanol
Formel:
C₁₀H₁₀N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159595 CAS-Nummer: 70817-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...