Ethyl 6-formylpyridine-2-carboxylate 95%

Lieferant: Apollo Scientific

OR953615-250MG
APOSOR953615-250MGEA 0 EUR
APOSOR953615-250MG
Ethyl 6-formylpyridine-2-carboxylate 95%
Ethyl 6-formylpyridine-2-carboxylate
Formel: C₉H₉NO₃
Molecular Weight: 179.173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501347
CAS-Nummer: 21908-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR