2-Chloroethanesulphonic acid sodium salt hydrate ≥98%

Lieferant: Apollo Scientific

Synonyme: Sodium-2-chloroethanesulphonate hydrate

OR95369-1KG
APOSOR95369-1KGEA 195 EUR
APOSOR95369-1KG
2-Chloroethanesulphonic acid sodium salt hydrate ≥98%
2-Chloroethanesulphonic acid sodium salt hydrate

Bitte beachten Use only outdoors or in a well-ventilated area. Avoid breathing dust/fumes. Wear protective gloves. Wash all exposed external body areas thoroughly after handling. If in eyes rinse cautiously with hazard eye protection and face protection.

Formel: C₂H₄ClNaO₃S.H₂O
Molecular Weight: 184,57 g/mol
Schmelzpunkt: 292 °C
MDL: MFCD00007531
CAS-Nummer: 1089719-72-1

Bestellen

Spezifikation Testergebnisse

Purity 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR