6-Methylisatin 95%
Lieferant: Apollo Scientific
Synonyme:
6-Methyl isatinic anhydride
OR953722-1G
APOSOR953722-1GEA
87
EUR
APOSOR953722-1G
6-Methylisatin 95%
6-Methylisatin
Formel:
C₉H₇NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01718832 CAS-Nummer: 1128-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...