2-Chloro-4-iodo-5-methylpyridine 95%

Lieferant: Apollo Scientific
Achtung

OR953758-250MG OR953758-1G
APOSOR953758-250MGEA 37.7 EUR
APOSOR953758-250MG APOSOR953758-1G
2-Chloro-4-iodo-5-methylpyridine 95%
2-Chloro-4-iodo-5-methylpyridine
Formel: C₆H₅ClIN
Lagertemperatur: Gefrierschrank
MDL: MFCD16610529
CAS-Nummer: 1197957-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR