2-Chloro-4-iodo-5-methylpyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR953758-250MG
OR953758-1G
APOSOR953758-250MGEA
37.7
EUR
APOSOR953758-250MG
APOSOR953758-1G
2-Chloro-4-iodo-5-methylpyridine 95%
2-Chloro-4-iodo-5-methylpyridine
Formel:
C₆H₅ClIN Lagertemperatur: Gefrierschrank |
MDL:
MFCD16610529 CAS-Nummer: 1197957-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...