Imidazo[1,2-a]pyridin-2-yl-acetic acid 95%

Lieferant: Apollo Scientific

OR953789-1G OR953789-250MG
APOSOR953789-1GEA 857 EUR
APOSOR953789-1G APOSOR953789-250MG
Imidazo[1,2-a]pyridin-2-yl-acetic acid 95%
Imidazo[1,2-a]pyridin-2-yl-acetic acid
Formel: C₉H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03434391
CAS-Nummer: 19741-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR