Ethyl imidazo[1,5-a]pyridine-1-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Imidazo[1,5-a]pyridine-1-carboxylic acid ethyl ester

OR953793-1G
APOSOR953793-1GEA 108 EUR
APOSOR953793-1G
Ethyl imidazo[1,5-a]pyridine-1-carboxylate 95%
Ethyl imidazo[1,5-a]pyridine-1-carboxylate
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,1986 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01237556
CAS-Nummer: 119448-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR