1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile 98%

Lieferant: Apollo Scientific

OR953815-5G
APOSOR953815-5GEA 215 EUR
APOSOR953815-5G
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile 98%
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile
Formel: C₈H₅N₃
Molecular Weight: 143.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659684
CAS-Nummer: 517918-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR