L-(-)-3-N-Boc-amino-2-azepanone 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-tert-Butyl (2-oxoazepan-3-yl)carbamate, tert-Butyl N-[(3S)-2-oxoazepan-3-yl]carbamate

OR953841-1G
APOSOR953841-1GEA 62.5 EUR
APOSOR953841-1G
L-(-)-3-N-Boc-amino-2-azepanone 96%
L-(-)-3-N-Boc-amino-2-azepanone
Formel: C₁₁H₂₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00798169
CAS-Nummer: 76944-95-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR