1-(2-Methoxy-phenyl)-ethylamine 95%

Lieferant: Apollo Scientific

Synonyme: 1-(2-Methoxyphenyl)ethanamine

OR953919-1G
APOSOR953919-1GEA 121 EUR
APOSOR953919-1G
1-(2-Methoxy-phenyl)-ethylamine 95%
1-(2-Methoxy-phenyl)-ethylamine
Formel: C₉H₁₃NO
Molecular Weight: 151,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00593475
CAS-Nummer: 40023-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR