Quinazoline-4-thiol 95%

Lieferant: Apollo Scientific

OR953937-1G
APOSOR953937-1GEA 632 EUR
APOSOR953937-1G
Quinazoline-4-thiol 95%
Quinazoline-4-thiol
Formel: C₈H₆N₂S
Molecular Weight: 162,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00091877
CAS-Nummer: 3337-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR